ContactUsLinkComponent
Middlebrook 7H9 Broth Base, CRITERION™, Hardy Diagnostics
Middlebrook 7H9 Broth Base, CRITERION™, Hardy Diagnostics
  89405-708
Product available on GSA Advantage®
 :  
undefined
Middlebrook 7H9 Broth Base, CRITERION™, Hardy Diagnostics
  89405-708
 :  C6301
Product available on GSA Advantage®
 :  
  • Pack type:
    Bottle, wide mouth
  • Size:
    500 g
  • Storage Temperature:
    –2-30°C
  • Cat. No.:
    89405-708
  • Supplier no.:
    C6301
CRITERION™ Middlebrook 7H9 Broth Base is primarily for the cultivation of mycobacteria
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
 : BSE Free