MacConkey Agar without Crystal Violet CRITERION™, Hardy Diagnostics
CRITERION™ MacConkey Agar without Crystal Violet is recommended for the detection of coliforms and enteric pathogens in water, waste water, and foods, as well as for differentiating Mycobacteria fortuitum-chelonei complex from other rapidly growing mycobacteria
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
: BSE Free
- Storage Temperature:–2-30°C
Frequently Bought Together