m Brilliant Green Broth, CRITERION™, Hardy Diagnostics
Catalog # 89406-594
Supplier: Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:
Specifications
- Pack type:Bottle, wide mouth
- Size:500 g
- Storage Temperature:–2-30°C
- Cat. No.:89406-594
- Supplier no.:C7941
Specifications
About this item
CRITERION™ m - Brilliant Green Broth is for the selective and differential isolation of Salmonella from water by membrane filtration
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free