Orders LinkContactUsLinkComponent
Indole Nitrate Medium, CRITERION™, Hardy Diagnostics
Indole Nitrate Medium, CRITERION™, Hardy Diagnostics
Indole Nitrate Medium, CRITERION™, Hardy Diagnostics
Supplier:  Hardy Diagnostics
Certificates
Indole Nitrate Medium, CRITERION™, Hardy Diagnostics
Indole Nitrate Medium, CRITERION™, Hardy Diagnostics
4 Options Available Below
Product Details & Documents
CRITERION™ Indole Nitrate Medium for the detection of indole production and nitrate reduction by microorganisms
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
Specifications for Product Family
Specifications
Specifications
  • Catalog No:
  • 89405-496
  • 89405-498
  • 89405-500
  • 89405-502
  • Storage Temperature:
  • –2-30°C
  • –2-30°C
  • –2-30°C
  • –2-30°C
Specifications
Product Family Options
Product Information
  • Size
    Final volume
    Pack type
    Availability
    Price
  • 122.4 g
    2 L
    Zip-pouch to make 2 L, Mylar®
    Log in for availability
    Log in for availability
    Each
    Each - $72.60
    $72.60
    Each
       $72.60
    Minimum order quantity is 1
    Supplier Number: C5870
    Specifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    89405-496
    Storage Temperature
    –2-30°C
    Product available on GSA Advantage®
  • 500 g
    Bottle, wide mouth
    Log in for availability
    Log in for availability
    Each
    Each - $245.00
    $245.00
    Each
       $245.00
    Minimum order quantity is 1
    Supplier Number: C5871
    Specifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    89405-498
    Storage Temperature
    –2-30°C
    Product available on GSA Advantage®
  • 2 kg
    Bucket
    Log in for availability
    Log in for availability
    Each
    Each - $964.00
    $964.00
    Each
       $964.00
    Minimum order quantity is 1
    Supplier Number: C5872
    Specifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    89405-500
    Storage Temperature
    –2-30°C
    Product available on GSA Advantage®
  • 10 kg
    Bucket
    Log in for availability
    Log in for availability
    Each
    Each - $4,450.00
    $4,450.00
    Each
       $4,450.00
    Minimum order quantity is 1
    Supplier Number: C5873
    Specifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    89405-502
    Storage Temperature
    –2-30°C
    Product available on GSA Advantage®