ContactUsLinkComponent
Hemoglobin Powder, CRITERION™, Hardy Diagnostics
  89405-490
Product available on GSA Advantage®
 :  
undefined
Hemoglobin Powder, CRITERION™, Hardy Diagnostics
  89405-490
 :  C5851
Product available on GSA Advantage®
 :  
  • Pack type:
    Bottle, wide mouth
  • Size:
    500 g
  • Storage Temperature:
    –2-30°C
  • Cat. No.:
    89405-490
  • Supplier no.:
    C5851
CRITERION™ Hemoglobin Powder is used as a supplement for GC Agar Base in the preparation of Chocolate Agar
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Chocolate Agar is used to cultivate fastidious organisms such as Neisseria spp. and Haemophilus spp. Dehydratedculture media with engineered container.
 : BSE Free