Columbia Agar Base, CRITERION™, Hardy Diagnostics
Supplier: Hardy Diagnostics
Certificates
About this item
CRITERION™ Columbia Agar Base is used for cultivating fastidious and non-fastidious microorganisms with or without the addition of blood
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
It is an enriched agar medium.
Certifications: BSE Free
Specifications
- Storage Temperature:–2-30°C