Campylobacter Agar Base, CRITERION™, Hardy Diagnostics
Supplier: Hardy Diagnostics
Certificates
About this item
CRITERION™ Campy Selective Agar Base is used for the selective isolation of Campylobacter jejuni and Campylobacter coli from human, animal, avian and environmental specimens
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Campylobacter Selective Supplement (Karmali), product #SR167E, is required for this medium. Dehydratedculture media with engineered container.
Certifications: BSE Free
Specifications
- Storage Temperature:–2-30°C
Specifications
Related Information
Certificates
Search by Lot Number Frequently Bought Together