BCSA Base, CRITERION™, Hardy Diagnostics
Supplier: Hardy Diagnostics
About this item
CRITERION™ BCSA Base, used for the selective isolation of Burkholderia (Pseudomonas) cepacia
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Antibiotics Polymixin B, Gentamicin, and Vancomycin required. See product label for details. Dehydratedculture media with engineered container.
: BSE Free
3 Options Available Below
Product Details & Documents
CRITERION™ BCSA Base, used for the selective isolation of Burkholderia (Pseudomonas) cepacia
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Antibiotics Polymixin B, Gentamicin, and Vancomycin required. See product label for details. Dehydratedculture media with engineered container.
: BSE Free
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 89406-812
- 89406-816
- 89406-818
- Storage Temperature:
- –2-30°C
- –2-30°C
- –2-30°C
Specifications
Product Family Options
Product Information
- SizeFinal volumePack typeAvailabilityPrice
- Supplier Number: C8880Specifications:Product available on GSA Advantage®
More
Specifications
Cat. No.89406-812Storage Temperature–2-30°CProduct available on GSA Advantage® - Supplier Number: C8882Specifications:Product available on GSA Advantage®
More
Specifications
Cat. No.89406-816Storage Temperature–2-30°CProduct available on GSA Advantage® - Supplier Number: C8883Specifications:Product available on GSA Advantage®
More
Specifications
Cat. No.89406-818Storage Temperature–2-30°CProduct available on GSA Advantage®