- Bottle Capacity:1 L
- Description:FTA-1 Aspirator
- Electrical:110-230V, 12V input
- Vacuum Range:–500mbar
- Dimensions:160 x 210 x 340 mm
- Weight:1.5 kg
- Cat. No.:89402-922
- Supplier no.:FTA-1
The aspirator with trap flask FTA-1 with a 1L trap flask is designed for the aspiration or removal of any remaining alcohol or buffer from microtest tube walls during DNA/RNA purification and other macromolecule re-precipitation techniques
- All in one system with integrated pump
- Trap flask volume 1L
- Fitted with hydrophobic microbiological filter
- Vacuum pressure of -500mbar
- Small, compact and easy to use
- Perfect for small volume aspiration
Compact, efficient, easy to use. Flexible for the user by offering a choice of either a single tip operation or a multi-tip option with 8 tips for use on standard PCR type plates. The integrated micro-biological filter is hydrophobic and designed to trap any bacteria, viruses or infected particles which may have been present in the aspirated solutions. The unit can stand directly inside a workstation.
Optional MA-8, 8 Channel Adapter for FTA-1 available. Adaptor kit includes tube adaptor, 8 channel aspiration tip, FTA-1 holder. Can be autoclaved.
An ideal personal tool for independent operation away from an inline lab vacuum supply.