Order Entry
Puerto Rico
ContactUsLinkComponent
MRI Narrow, Lockable Carts with Roll Top Door, Polyethylene, TrippNT
MRI Narrow, Lockable Carts with Roll Top Door, Polyethylene, TrippNT
Catalog # 89408-452
Supplier:  TrippNT
undefined
MRI Narrow, Lockable Carts with Roll Top Door, Polyethylene, TrippNT
Catalog # 89408-452
Supplier:  TrippNT
Supplier Number:  50729

Specifications

  • Color:
    White with White Door
  • Description:
    MRI Narrow, Lockable Cart with Roll Top Door
  • Cat. No.:
    89408-452
  • Weight:
    39 lbs.
  • Supplier no.:
    50729

Specifications

About this item

Narrow, White Polyethylene MRI Lockable Cart with Roll Top (Tambour Door) is classified as MR Conditional and features 6 removable, one pocket drawers and an easy-open roll top door.

  • Narrow space saving design
  • Non-ferrous MRI locking cart
  • 6 removable trays
  • Easy opening locking tambour door
  • Chemically resistant polyethylene construction
  • Top has built in handle making it smooth and easy to maneuver
  • 3 inch casters
  • Fully assembled, ready to use out of the box
  • American made by TrippNT
  • Total Replacement Lifetime Guarantee

Roll top door is space saving and disappears into cart when open.