Order Entry
Puerto Rico
ContactUsLinkComponent
Corning® Bovine Pituitary Extract (BPE), Corning
Corning® Bovine Pituitary Extract (BPE), Corning
Catalog # 47743-648
Supplier:  Corning
   Low Manufacturing Impact
CAS Number:  
Corning® Bovine Pituitary Extract (BPE), Corning
Catalog # 47743-648
Supplier:  Corning
Supplier Number:  354123
   Low Manufacturing Impact
CAS Number:  

Specifications

  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental Certification:
    ISO 14001 - Bedford (reg. no. 14.4942)
  • Regulatory Status:
    RUO
  • Cat. No.:
    47743-648
  • Source:
    Bovine pituitary glands
  • Shelf life:
    790 days
  • Type:
    Media additive
  • Volume:
    15 mg
  • Supplier no.:
    354123

Specifications

About this item

BPE is a broadly used supplement to culture a variety of epithelial and endothelial cells

BPE contains growth factors (e.g., basic FGF) and hormones.

Tested for ability to promote proliferation of Fetal Bovine Heart Endothelial Cells (FBHEC) in medium containing 10% newborn calf serum. Filtered (0.2 µm membrane) and tested and found negative for the presence of bacteria, fungi, and mycoplasma.

BPE contains growth factors (e.g., basic FGF) and hormones.