Order Entry
Puerto Rico
ContactUsLinkComponent
NUCLEAR-ID® ≥93% (by HPLC), CELLESTIAL® red DNA stain
NUCLEAR-ID® ≥93% (by HPLC), CELLESTIAL® red DNA stain
Catalog # 89166-036
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
NUCLEAR-ID® ≥93% (by HPLC), CELLESTIAL® red DNA stain
Catalog # 89166-036
Supplier:  Enzo Life Sciences
Supplier Number:  ENZ-52406
CAS Number:  

Specifications

  • Size:
    200 µl
  • Cat. No.:
    89166-036
  • Supplier no.:
    52406

Specifications

Specification Test Results

  • Excitation maximum::
    566nm
  • Emission maximum::
    650nm

Specification Test Results

About this item

 :
Storage Temperature:Freezer