Order Entry
Puerto Rico
ContactUsLinkComponent
Di-2-ANEPEQ (JPW 1114) ≥90% (by HPLC), CELLESTIAL® membrane stain, for histology
Di-2-ANEPEQ (JPW 1114) ≥90% (by HPLC), CELLESTIAL® membrane stain, for histology
Catalog # 89165-994
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Di-2-ANEPEQ (JPW 1114) ≥90% (by HPLC), CELLESTIAL® membrane stain, for histology
Catalog # 89165-994
Supplier:  Enzo Life Sciences
Supplier Number:  ENZ-52201
CAS Number:  

Specifications

  • Size:
    5 mg
  • Cat. No.:
    89165-994
  • Supplier no.:
    52201

Specifications

About this item

 :
 :549.384 g/mol
Storage Temperature:Freezer
Formula:C₂₆H₃₅Br₂N₃