Order Entry
Puerto Rico
ContactUsLinkComponent
TSQ ≥98% (by HPLC), Ultrapure
TSQ ≥98% (by HPLC), Ultrapure
Catalog # 89165-984
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
TSQ ≥98% (by HPLC), Ultrapure
Catalog # 89165-984
Supplier:  Enzo Life Sciences
Supplier Number:  ENZ-52153
CAS Number:  

Specifications

  • Size:
    25 mg
  • Cat. No.:
    89165-984
  • Supplier no.:
    52153

Specifications

About this item

 :
 :328.38 g/mol
Storage Temperature:Freezer
CAS Number:109628-27-5
Formula:C₁₇H₁₆N₂O₃S