Order Entry
Puerto Rico
ContactUsLinkComponent
5(6)-Carboxy-2′,7′- dichlorofluorescein diacetate ≥98% (by HPLC), Ultrapure
5(6)-Carboxy-2′,7′- dichlorofluorescein diacetate ≥98% (by HPLC), Ultrapure
Catalog # 89165-976
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
5(6)-Carboxy-2′,7′- dichlorofluorescein diacetate ≥98% (by HPLC), Ultrapure
Catalog # 89165-976
Supplier:  Enzo Life Sciences
Supplier Number:  ENZ-52103
CAS Number:  

Specifications

  • Size:
    100 mg
  • Cat. No.:
    89165-976
  • Supplier no.:
    52103

Specifications

About this item

 :
 :529.28 g/mol
Storage Temperature:Freezer
CAS Number:127770-45-0
Formula:C₂₅H₁₄Cl₂O₉
MDL Number:MFCD08459585