$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Fura 2-AM
Fura 2-AM ≥95% (by HPLC), Ultrapure
Fura 2-AM ≥95% (by HPLC), Ultrapure
89165-942
:
Enzo Life Sciences
:
Fura 2-AM ≥95% (by HPLC), Ultrapure
89165-942
:
Enzo Life Sciences
:
ENZ-52006
:
Size:
1 mg
Cat. No.:
89165-942
Supplier no.:
52006
:
:
1001.857 g/mol
:
Freezer
:
108964-32-5
:
C₄₄H₄₇N₃O₂₄
:
MFCD00036976