To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Size:1 Kit
- Storage Conditions:–20 °C (Long Term)
- Application:Fluorescence Microscopy, HTS
- Cat. No.:89165-870
- Supplier no.:51004
Red Hydrogen Peroxide Assay Kit provides a simple absorbance-based or fluorometric assay to detect and quantify hydrogen peroxide or peroxidase activity in biochemical assays, cell extracts and in live cells
- Homogenous assay format, fully compatible with HTS automation
- Detect as little as 10 picomoles of hydrogen peroxide
- Mix and read reagents, with minimal hands-on time
It can also be used to detect a variety of oxidase activities through enzyme-coupled reactions. The kit is an optimized ‘mix and read’ assay that is suitable for both 96- and 348-well formats and is compatible with HTS liquid handling.