Order Entry
Puerto Rico
ContactUsLinkComponent
Aqua 431 dUTP, Enzo Life Sciences
Aqua 431 dUTP, Enzo Life Sciences
Catalog # 89165-856
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Aqua 431 dUTP, Enzo Life Sciences
Catalog # 89165-856
Supplier:  Enzo Life Sciences
Supplier Number:  ENZ-42853
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Concentration:
    1 mM
  • Description:
    Aqua 431 dUTP
  • Size:
    25 nmoL
  • Purity Detail:
    Purified by ion-exchange chromatography.
  • Purity:
    ≥93% (HPLC)
  • Formulation:
    Liquid. Solution in water.
  • Cat. No.:
    89165-856
  • Supplier no.:
    42853

Specifications

About this item

Aqua 431 [DEAC] dUTP is a coumarin derivative that can replace TTP in reactions in which it serves as a substrate for E. coli DNA polymerase (holoenzyme and Klenow fragment), T4 and Taq DNA polymerases, reverse transcriptase (from AMV and M-MuLV) and terminal transferase.

Fluorescently labelled probes can be prepared with this fluorescent nucleotide by a variety of methods including nick translation, random prime labelling, cDNA labelling and 3’-end labelling. Probes generated by these methods are suitable for use for the identification of specific sequences by in situ hybridization procedures on fixed cells and tissues by direct fluorescence detection. Aqua 431 dUTP can also be used for multicolor fluorescence labelling.