BIO-PROBE® Cytomegalovirus Probe, Enzo Life Sciences
89165-724
:
- Description:BIO-PROBE® Cytomegalovirus Probe
- Size:2 µg
- Concentration:20µg/ml
- Formulation:Liquid. In 10mM TRIS HCl, pH 7.5, containing 1mM EDTA.
- Cat. No.:89165-724
- Supplier no.:40835
The Cytomegalovirus (CMV) BioProbe® labeled probe is prepared by nick translation of cloned fragments of the CMV Towne strain.
The fragments include a total of 30-31 kb of DNA, approximately 20% of the CMV genome. The probe is specific for cytomegalovirus DNA. It does not hybridize to DNAs of other herpesviruses (i.e., herpes simplex virus or Epstein-Barr virus) but it has been found to hybridize to the DNA of CMV clinical isolates. Fragment size range: 100-1000 base pairs (as estimated by agarose gel electrophoresis).