Order Entry
United States
ContactUsLinkComponent
 
Anti-VIM Mouse Monoclonal Antibody
  89165-480
undefined
Anti-VIM Mouse Monoclonal Antibody
  89165-480
 :  VG6100-0100
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Vimentin
  • Antigen Symbol:
    VIM
  • Clonality:
    Monoclonal
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 µL
  • Environmentally Preferable:
  • Antigen Synonyms:
    CTRCT30|HEL113
  • Cat. No.:
    89165-480
  • Supplier no.:
    VG6100-0100

 

 

Vimentin is a the type III intermediate filament (IF) protein expressed in a variety of mesenchymal and non-mesenchymal cells, including muscle cells, astrocytes, neurons, endothelial cells and fibroblasts. During development of the mammalian nervous system vimentin is transiently expressed in neuronal precursor cells, often with the type VI IF, nestin. Vimentin appears to play a role in many intracellular processes, recent examples from the literature include adhesion, glial scar formation, neuronal migration, and the normal development of Bergmann glia.

  • For Immunohistochemistry, Western Blot

Recognizes human Vimentin.

Type: Primary
Antigen: VIM
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human