Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant UBCH6 (from E. coli), HIS Tag
Human Recombinant UBCH6 (from E. coli), HIS Tag
Catalog # 89165-278
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Human Recombinant UBCH6 (from E. coli), HIS Tag
Catalog # 89165-278
Supplier:  Enzo Life Sciences
Supplier Number:  UW8710-0100
CAS Number:  

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    100 µg
  • Tag sequence:
    HIS Tag
  • Protein/Peptide Name:
    UBCH6
  • Purity:
    >95% (SDS-PAGE)
  • Concentration:
    1.7mg/ml
  • Formulation:
    Liquid. In PBS, pH 7.2, containing 1mM DTT and 10% glycerol.
  • Cat. No.:
    89165-278
  • Supplier no.:
    UW8710-0100

Specifications

About this item

Produced in E. coli.

Ubiquitinylation of proteins constitutes an important cellular mechanism for targeting short-lived proteins for degradation by the 26S proteasome. Three classes of enzymes are involved in the conjugation of ubiquitin to proteins. E1, the ubiquitin activating enzyme, activates ubiquitin through the ATP-dependent formation of a high-energy thiol ester bond between the carboxyl terminus of ubiquitin and the active-site cysteine within E1. This E1-activated ubiquitin is transferred to a cysteine residue of an E2, or ubiquitin-conjugating enzyme (UbC). E2 enzymes, either by themselves or in conjunction with E3 enzymes (ubiquitin ligases), then transfer ubiquitin to target proteins forming stable isopeptide bonds resulting in multi-ubiquitin chain formation. It is the diverse combinations of E2-E3 complexes that are thought to define substrate specificity. Sequence comparison reveals that UbcH6 is a novel member of an evolutionally conserved subfamily of E2s that includes UbcH5 and the S. cerevisiae derived Ubc4. However, UbcH6 appears to be unique among these E2s since it contains an amino-terminal extension of approximately 40 amino acids, the functional significance of which extension is presently unknown. Confusingly, UbcH6 it is not related to the S. cerevisiae derived Ubc6. Regardless of amino acid sequence homologies, it appears that UbcH6 may be structurally more similar to UbcH5a than UbcH5a is itself to either UbcH5b or UbcH5c. Despite these sequence similarities, UbcH6 does not efficiently substitute for UbcH5 or S. cerevisiae derived Ubc4 in E6-AP-dependent protein ubiquitinylation or many other HECT-domain mediated events. The real function of UbcH6 appears yet to be fully defined although a recent publication suggests that UbcH6 plays a role in the ubiquitinylation of human IAPs (inhibitors of apoptosis).