Order Entry
Puerto Rico
ContactUsLinkComponent
Dihydrosphingosine-1-phosphate, D-erythro ≥98%
Dihydrosphingosine-1-phosphate, D-erythro ≥98%
Catalog # 89164-612
Supplier:  Enzo Life Sciences
CAS Number:  
Dihydrosphingosine-1-phosphate, D-erythro ≥98%
Catalog # 89164-612
Supplier:  Enzo Life Sciences
Supplier Number:  BML-SL143-0001
CAS Number:  

Specifications

  • Size:
    1 mg
  • Cat. No.:
    89164-612
  • Supplier no.:
    SL143-0001

Specifications

Specification Test Results

  • Purity:
    ≥98%

Specification Test Results

About this item

 :
 :381.5 g/mol
Storage Temperature:Freezer
CAS Number:28508-07-8
Formula:C₁₈H₄₀NO₅P
MDL Number:MFCD00871363