- Protein/Peptide Type:Recombinant
- Species:Human
- Size:10 µg
- Protein/Peptide Name:MMP-14 (catalytic domain) (112-298)
- Cat. No.:89163-908
- Supplier no.:SE259-0010
Produced in E.coli. Active Matrix Metalloproteinase-14 (MMP-14, Membrane-Type Matrix Metalloproteinase 1, MT1-MMP) catalytic domain from human cDNA. The enzyme consists of the catalytic domain of human MMP-14 (Tyr112-Arg298, NM_004995) with a C-terminal purification tag. This represents a naturally-occurring active form of MMP-14 which lacks the C-terminal hemopexin domain. MMPs lacking this domain cannot cleave native collagens; however, activity toward other targets such as gelatin, casein, or peptide substrates is unaffected.