Anti-14-3-3 Rabbit Polyclonal Antibody
Supplier: Enzo Life Sciences
Certificates
About this item
Host: Rabbit
- For Immunohistochemistry, Immunoprecipitation, Western Blot
Recognizes mammalian, chicken, plants, yeast and invertebrate 14-3-3 pan specific.
Type: Primary
Antigen: 14-3-3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mammals
Specifications
- Catalog No:
- 89163-172
- Antigen Symbol:
- 14-3-3
- Antigen Name:
- 14-3-3 protein
- Antigen Synonyms:
- beta isoform|14-3-3 alpha|Protein 1054|Alpha polypeptide|Protein kinase C inhibitor protein 1|Tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein|protein kinase C inhibitor protein-1|14-3-3 Protein beta/alpha|brain Protein 14-3-3
- Antibody Type:
- Primary
- Clonality:
- Polyclonal
- Reactivity:
- Mammals
- Host:
- Rabbit
- Western Blot:
- Yes
- ImmunoChemistry:
- Yes
- ImmunoPrecipitation:
- Yes