Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-IP3 Receptor TYPE 1 Rabbit Polyclonal Antibody
Anti-IP3 Receptor TYPE 1 Rabbit Polyclonal Antibody
Catalog # 89162-788
Supplier:  Enzo Life Sciences
undefined
Anti-IP3 Receptor TYPE 1 Rabbit Polyclonal Antibody
Catalog # 89162-788
Supplier:  Enzo Life Sciences
Supplier Number:  BML-SA254-0100

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    IP3 Receptor TYPE 1
  • Antigen Symbol:
    IP3 Receptor TYPE 1
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 µL
  • Form:
    Liquid. Neat serum containing 0.05% sodium azide.
  • Cat. No.:
    89162-788
  • Supplier no.:
    SA254-0100

Specifications

About this item

Host: Rabbit

  • For Immunohistochemistry, Western Blot

Recognizes human, rat and dog IP3R1. Detects a band of ~240kDa by Western blot.

Type: Primary
Antigen: IP3 RECEPTOR TYPE 1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human