Anti-Cop9 Signalosome Csn7 Subunit Rabbit Polyclonal Antibody
Supplier: Enzo Life Sciences
Certificates
About this item
Host: Rabbit
- For Immunoprecipitation, Western Blot
Recognizes human COP9 signalosome (Csn7 subunit).
Type: Primary
Antigen: COP9 SIGNALOSOME CSN7 SUBUNIT
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 89162-128
- 89162-130
- Antigen Symbol:
- Cop9 Signalosome Csn7 Subunit
- Cop9 Signalosome Csn7 Subunit
- Antigen Name:
- Cop9 Signalosome Csn7 Subunit
- Cop9 Signalosome Csn7 Subunit
- Antibody Type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Western Blot:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Form:
- Liquid. In PBS containing 10mM sodium azide.