Order Entry
Puerto Rico
ContactUsLinkComponent
Epoxomicin ≥95% (by HPLC)
Epoxomicin ≥95% (by HPLC)
Catalog # 89161-634
Supplier:  Enzo Life Sciences
CAS Number:  
Epoxomicin ≥95% (by HPLC)
Catalog # 89161-634
Supplier:  Enzo Life Sciences
Supplier Number:  BML-PI127-0100
CAS Number:  

Specifications

  • Size:
    100 µg
  • Cat. No.:
    89161-634
  • Supplier no.:
    PI127-0100

Specifications

About this item

 :
 :554.7 g/mol
CAS Number:134381-21-8
Formula:C₂₈H₅₀N₄O₇
MDL Number:MFCD03791061