Order Entry
Puerto Rico
ContactUsLinkComponent
L-α-Phosphatidylinositol-3,4,5-P3 ·7Na ≥98% (by TLC)
Irritant
L-α-Phosphatidylinositol-3,4,5-P3 ·7Na ≥98% (by TLC)
Catalog # 89161-556
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Irritant
L-α-Phosphatidylinositol-3,4,5-P3 ·7Na ≥98% (by TLC)
Catalog # 89161-556
Supplier:  Enzo Life Sciences
Supplier Number:  BML-PH107-0100
CAS Number:  

Specifications

  • Size:
    100 µg
  • Cat. No.:
    89161-556
  • Supplier no.:
    PH107-0100

Specifications

Specification Test Results

  • Purity:
    ≥98% (TLC)
  • Formulation:
    Lyophilised

Specification Test Results

About this item

 :
 :1204.8 g/mol
 :GHS07
Storage Temperature:Freezer
 :383907-36-6
Formula:C41H75O22P4·7Na