Order Entry
Puerto Rico
ContactUsLinkComponent
8-Methoxymethyl-IBMX ≥98% (by TLC)
8-Methoxymethyl-IBMX ≥98% (by TLC)
Catalog # 89161-350
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
8-Methoxymethyl-IBMX ≥98% (by TLC)
Catalog # 89161-350
Supplier:  Enzo Life Sciences
Supplier Number:  BML-PD150-0010
CAS Number:  

Specifications

  • Size:
    10 mg
  • Cat. No.:
    89161-350
  • Supplier no.:
    PD150-0010

Specifications

About this item

 :
 :266.3 g/mol
Storage Temperature:Ambient
CAS Number:78033-08-6
Formula:C₁₂H₁₈N₄O₃