H-Arg-AMC·2HCl
Catalog # 89160-982
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Size:25 mg
- Cat. No.:89160-982
- Supplier no.:P135-0025
Specifications
About this item
:
:404.3 g/mol
CAS Number:113712-08-6
Formula:C₁₆H₂₁N₅O₃·2HCl
MDL Number:MFCD00155578