Order Entry
Puerto Rico
ContactUsLinkComponent
Autocamtide-2
Autocamtide-2
Catalog # 89160-922
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Autocamtide-2
Catalog # 89160-922
Supplier:  Enzo Life Sciences
Supplier Number:  BML-P101-0001
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Size:
    1 mg
  • Protein/Peptide Name:
    Autocamtide-2
  • Purity:
    >95% (HPLC)
  • Formulation:
    Lyophilized
  • Cat. No.:
    89160-922
  • Supplier no.:
    P101-0001

Specifications

About this item

CaM kinase inhibitor

Sequence, KKALRRQETVDAL, derived from Ca2+/calmodulin-dependent protein kinase II autophosphorylation site RQETVD plus basic residues to facilitate phosphocellulose binding. Contains consensus phosphorylation sequence R-X-X-S/T. Highly selective CaMKII substrate (Km=2 µM for CaMKII; 50 µM for PKC; no activity towards PKA).