Order Entry
Puerto Rico
ContactUsLinkComponent
Melatonin ≥98% (by HPLC)
Melatonin ≥98% (by HPLC)
Catalog # 89160-870
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Melatonin ≥98% (by HPLC)
Catalog # 89160-870
Supplier:  Enzo Life Sciences
Supplier Number:  BML-NS520-0001
CAS Number:  

Specifications

  • Size:
    1 g
  • Cat. No.:
    89160-870
  • Supplier no.:
    NS520-0001

Specifications

About this item

 :
 :232.28 g/mol
Storage Temperature:Freezer
CAS Number:73-31-4
Formula:C₁₃H₁₆N₂O₂
MDL Number:MFCD00005655