Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
FLUOR DE LYS® Deacetylated Standard, Enzo Life Sciences
FLUOR DE LYS® Deacetylated Standard, Enzo Life Sciences
FLUOR DE LYS® Deacetylated Standard, Enzo Life Sciences
Catalog # 89160-324
Supplier:
Enzo Life Sciences
CAS Number:
Expand image
FLUOR DE LYS® Deacetylated Standard, Enzo Life Sciences
Catalog # 89160-324
Supplier:
Enzo Life Sciences
Supplier Number:
BML-KI142-0030
CAS Number:
Specifications
Description:
Fluor-de-Lys® Deacetylated Standard
Size:
30 µl
Cat. No.:
89160-324
Supplier no.:
KI142-0030
Specifications
About this item
Product Details
Product Details