cis-5,8,11-Eicosatrienoic acid ≥97% (by HPLC), colourless oil
Supplier: Enzo Life Sciences
Certificates
About this item
CAS Number:20590-32-3
:
Formula:C₂₀H₃₄O₂
MDL Number:MFCD00065719
:306.5 g/mol
Storage Temperature:Freezer
Specification Test Results
- Purity:≥97% (HPLC)