Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
DuP-697
DuP-697 ≥98% (by TLC)
DuP-697 ≥98% (by TLC)
DuP-697 ≥98% (by TLC)
Supplier:
Enzo Life Sciences
Certificates
Expand image
DuP-697 ≥98% (by TLC)
Supplier:
Enzo Life Sciences
About this item
CAS Number:
88149-94-4
:
Formula:
C₁₇H₁₂BrFO₂S₂
:
411.3 g/mol