Anti-Met Enkephalin Rabbit Polyclonal Antibody
Supplier: Enzo Life Sciences
Certificates
About this item
Host: Rabbit
- For Immunohistochemistry
Recognizes methionine enkephalin in a wide range of species.
Type: Primary
Antigen: MET ENKEPHALIN
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Species Independent
Specifications
- Catalog No:
- 89157-440
- 89157-436
- Antigen Symbol:
- Met Enkephalin
- Met Enkephalin
- Antigen Name:
- Met Enkephalin
- Met Enkephalin
- Antibody Type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Reactivity:
- Multiple
- Multiple
- Host:
- Rabbit
- Rabbit
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid. Antiserum containing 0.1% sodium azide.