Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
L-Mimosine
L-Mimosine ≥98%
L-Mimosine ≥98%
89156-898
:
Enzo Life Sciences
:
L-Mimosine ≥98%
89156-898
:
Enzo Life Sciences
:
BML-CC102-0500
:
Size:
500 mg
Cat. No.:
89156-898
Supplier no.:
CC102-0500
:
:
198.2 g/mol
:
Ambient
:
500-44-7
:
C₈H₁₀N₂O₄
:
MFCD00075909