Order Entry
Puerto Rico
ContactUsLinkComponent
Calcimycin (A23187) (from Streptomyces chartreusis) ≥99% (by TLC)
Calcimycin (A23187) (from Streptomyces chartreusis) ≥99% (by TLC)
Catalog # 89156-726
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Calcimycin (A23187) (from Streptomyces chartreusis) ≥99% (by TLC)
Catalog # 89156-726
Supplier:  Enzo Life Sciences
Supplier Number:  BML-CA100-0010
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    10 mg
  • Cat. No.:
    89156-726
  • Supplier no.:
    CA100-0010

Specifications

About this item

 :
 :523.63 g/mol
Storage Temperature:Freezer
CAS Number:52665-69-7
Formula:C₂₉H₃₇N₃O₆
MDL Number:MFCD00151202