
(±)Epibatidine ≥99% (by HPLC)
Catalog # 89156-712
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Size:5 mg
- Cat. No.:89156-712
- Supplier no.:C114-0005
Specifications
Specification Test Results
- Purity:≥99% (HPLC)
Specification Test Results
About this item
:
:208.7 g/mol
UN:2811
ADR:6.1,I
:GHS06
CAS Number:140111-52-0
Formula:C₁₁H₁₃ClN₂