To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Size:96 Wells
- Storage Conditions:–20 °C (Long Term)
- Application:Fluorescent Detection, HTS Activity Assay
- Cat. No.:89156-546
- Supplier no.:AK740-0001
A QuantiZyme™ Assay System
The Suc-LLVY-AMC fluorogenic peptide substrate supplied with this kit is a proteasome substrate commonly used for measuring chymotrypsin-like peptidase activity. The detection of proteolytic activity is based on the release of free AMC fluorophore and offers the following advantages: non-radioactive, convenient real-time kinetics of substrate cleavage, and excellent sensitivity. A proteasome inhibitor, epoxomicin, is included as a test inhibitor for screening purposes.
Fluorogenic, non-radioactive assay designed to measure chymotrypsin-like protease activity of purified 20S proteasome. This 96-well assay is useful for screening inhibitors and modulators of the 20S proteasome.