Order Entry
Puerto Rico
ContactUsLinkComponent
ALDetect™ Lipid Peroxidation Assay Kit, Enzo Life Sciences
ALDetect™ Lipid Peroxidation Assay Kit, Enzo Life Sciences
Catalog # 89156-446
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
ALDetect™ Lipid Peroxidation Assay Kit, Enzo Life Sciences
Catalog # 89156-446
Supplier:  Enzo Life Sciences
Supplier Number:  BML-AK170-0001
CAS Number:  

Specifications

  • Size:
    100 Tests
  • Storage Conditions:
    4 °C (Long Term)
  • Application:
    Colorimetric Detection
  • Cat. No.:
    89156-446
  • Supplier no.:
    AK170-0001

Specifications

About this item

The assay is designed to measure either MDA alone (in hydrochloric acid) or MDA in combination with 4-hydroxyalkenals (in methanesulfonic acid). It is based on the reaction of a chromogenic reagent, N-methyl-2-phenylindole with MDA and 4-hydroxyalkenals to yield a stable chromophore with maximal absorbance at 586 nm.

Lipid peroxidation is a well-established mechanism of cellular injury in both plants and animals, and is used as an indicator of oxidative stress in cells and tissues

Polyunsaturated fatty acid peroxides generate malondialdehyde (MDA) and 4-hydroxyalkenals (HAE) upon decomposition. Measurement of malondialdehyde and 4-hydroxyalkenals has been used as an indicator of lipid peroxidation.

Lipid peroxides are unstable and decompose to form a complex series of compounds including reactive aldehydes.

Delivery information: Kit includes Reagent R1: N-methyl-2-phenylindole, in acetonitrile (3x18 mL), Reagent R2: conc. methanesulfonic acid (16.5 mL), MDA Standard: 1,1,3,3-tetramethoxypropane (TMOP) in TRIS-hydrochloric acid (1 mL), Diluent: ferric iron in methanol (30 mL).