Anti-RANKL Rat Monoclonal Antibody
Catalog # 89155-084
Supplier: Enzo Life Sciences
Specifications
- Antibody Type:Primary
- Antigen Name:Receptor activator of nuclear factor kappa-B ligand
- Antigen Symbol:RANKL
- Clonality:Monoclonal
- ELISA:Yes
- Host:Rat
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid. In PBS containing 0.02% sodium azide.
- Antigen Synonyms:RANKLigand|Osteoprotegerin ligand|OPGL|sOdf|ODF|TRANCE|CD254|TNF-related activation-induced cytokine|RANKL|Receptor activator of nuclear factor kappa B ligand|hRANKL2|OPTB2
- Concentration:1mg/ml
- Purification:≥95% (SDS-PAGE)
- Cat. No.:89155-084
- Supplier no.:804-830-C100
Specifications
About this item
Host: Rat, Isotype: IgG1
- For ELISA, Flow Cytometry, Immunohistochemistry, Western Blot
Recognizes human RANKL.
Type: Primary
Antigen: RANKL
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human