Order Entry
United States
ContactUsLinkComponent
Anti-TRAIL-R1 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-TRAIL-R1 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 89154-002
Supplier:  Avantor
undefined
Anti-TRAIL-R1 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 89154-002
Supplier:  Avantor
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    TNF receptor superfamily member 10a
  • Antigen Symbol:
    TRAIL-R1
  • Clonality:
    Monoclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Size:
    100 Tests
  • Environmentally Preferable:
  • Form:
    Liquid. 50µg in PBS containing 0.09% sodium azide.
  • Antigen Synonyms:
    TRAILR1|DR4|APO2|CD261|TRAILR-1|MGC9365
  • Concentration:
    0.5mg/ml
  • Purification:
    ≥95%
  • Cat. No.:
    89154-002
  • Supplier no.:
    804297FT100

Specifications

About this item

Host: Mouse, Isotype: IgG1

  • For Immunocytochemistry

Recognizes human TRAIL-R1. Does not cross-react with human TRAIL-R2, -R3 or -R4.

Type: Primary
Antigen: TRAIL-R1
Clonality: Monoclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human