Anti-BAFF Rat Monoclonal Antibody (Biotin)
Catalog # 89153-772
Supplier: Enzo Life Sciences
Specifications
- Antibody Type:Primary
- Antigen Name:B-cell activating factor
- Antigen Symbol:BAFF
- Clonality:Monoclonal
- Conjugation:Biotin
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Rat
- Isotype:IgG1
- Reactivity:Mouse
- Size:100 µg
- Form:Liquid. In PBS containing 0.02% sodium azide.
- Antigen Synonyms:CD257|ZTNF4|THANK|TALL-1|TNFSF20|DTL|TNFSF13B|TALL1|TNLG7A|BLYS
- Concentration:1mg/ml
- Purification:95% (SDS-PAGE).
- Cat. No.:89153-772
- Supplier no.:804157BC100
Specifications
About this item
Host: Rat, Isotype: IgG1
- For ELISA, Flow Cytometry
Recognizes mouse BAFF.
Type: Primary
Antigen: BAFF
Clonality: Monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse