Galantamine hydrobromide ≥98% (by HPLC)
Catalog # 89152-546
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Size:50 mg
- Cat. No.:89152-546
- Supplier no.:550-336-M050
Specifications
About this item
:
:368.27 g/mol
Storage Temperature:Freezer
CAS Number:1953-04-4
Formula:C₁₇H₂₁NO₃·HBr
MDL Number:MFCD00067672