Order Entry
Puerto Rico
ContactUsLinkComponent
Procainamide hydrochloride ≥98% (by HPLC)
Irritant
Procainamide hydrochloride ≥98% (by HPLC)
Catalog # 89152-474
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Irritant
Procainamide hydrochloride ≥98% (by HPLC)
Catalog # 89152-474
Supplier:  Enzo Life Sciences
Supplier Number:  550-287-G010
CAS Number:  

Specifications

  • Size:
    10 g
  • Cat. No.:
    89152-474
  • Supplier no.:
    550-287-G010

Specifications

Specification Test Results

  • Purity (HPLC):
    ≥98%

Specification Test Results

About this item

 :
 :271.789 g/mol
Merck Index:14,07756
EINECS:210-381-7
 :GHS07
Storage Temperature:Ambient
CAS Number:614-39-1
Formula:C₁₃H₂₂ClN₃O
MDL Number:MFCD00012998