Order Entry
Puerto Rico
ContactUsLinkComponent
Lidocaine hydrochloride hydrate ≥98%
Lidocaine hydrochloride hydrate ≥98%
Catalog # 89152-398
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Lidocaine hydrochloride hydrate ≥98%
Catalog # 89152-398
Supplier:  Enzo Life Sciences
Supplier Number:  550-251-G005
CAS Number:  

Specifications

  • Size:
    5 g
  • Cat. No.:
    89152-398
  • Supplier no.:
    550-251-G005

Specifications

About this item

 :
 :270.8 g/mol (anhy)
CAS Number:6108-05-0
Formula:C₁₄H₂₃ClN₂O·xH₂O