Order Entry
Puerto Rico
ContactUsLinkComponent
Pilocarpine hydrochloride ≥98% (by titrimetric analysis)
Pilocarpine hydrochloride ≥98% (by titrimetric analysis)
Catalog # 89152-144
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Pilocarpine hydrochloride ≥98% (by titrimetric analysis)
Catalog # 89152-144
Supplier:  Enzo Life Sciences
Supplier Number:  550-092-G001
CAS Number:  

Specifications

  • Size:
    1 g
  • Cat. No.:
    89152-144
  • Supplier no.:
    550-092-G001

Specifications

About this item

 :
CAS Number:54-71-7
Formula:C₁₁H₁₇ClN₂O₂
MDL Number:MFCD00012722