Order Entry
Puerto Rico
ContactUsLinkComponent
L-Noradrenaline bitartrate monohydrate ≥98%
 
 
L-Noradrenaline bitartrate monohydrate ≥98%
  89152-122
 :  
undefined
 
 
L-Noradrenaline bitartrate monohydrate ≥98%
  89152-122
 :  550-076-G001
 :  

 

  • Size:
    1 g
  • Cat. No.:
    89152-122
  • Supplier no.:
    550-076-G001

 

 

  • Purity (assay):
    ≥98%

 

 

 :
 :319.27 g/mol
 :14,06695
 :Refrigerator
 :2811
 :6.1,II
 :GHS06
 :GHS08
 :108341-18-0
 :C₈H₁₁NO₃
 :MFCD00036384