Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant IL-35:Fc
Human Recombinant IL-35:Fc
Catalog # 89151-952
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Human Recombinant IL-35:Fc
Catalog # 89151-952
Supplier:  Enzo Life Sciences
Supplier Number:  522-140-C010
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Species:
    Human
  • Size:
    10 µg
  • Endotoxin Content:
    <0.1EU/µg purified protein (LAL test; Lonza).
  • Biological Activity:
    Inhibits human mononuclear cell proliferation.
  • Reconstitution Instructions:
    Reconstitute with 100µl sterile water. Further dilutions should be made with medium containing 5% fetal calf serum or a carrier protein.
  • Protein/Peptide Name:
    IL-35:Fc
  • Purity:
    >95% (SDS-PAGE)
  • Concentration:
    0.1mg/ml after reconstitution.
  • Formulation:
    Lyophilized. Contains PBS.
  • Cat. No.:
    89151-952
  • Supplier no.:
    522-140-C010

Specifications

About this item

Produced in HEK 293 cells. Human EBI3 (aa 21-229) linked to human IL-12A (aa 23-219) via the linker GGGSGGGSGGGSGGGS is fused at the C-terminus to the Fc portion of human IgG1 and a FLAG®-tag.

EBI3 (Epstein-Barr virus-induced gene 3 protein; EBV-induced gene 3 protein; IL-27B; IL-27 subunit β) and IL-12A (IL-12 subunit p35; CLMF p35) form the heterodimeric cytokine IL-35 (interleukin-35) involved in T cell regulation. IL-35 is highly expressed in regulatory T cell (Treg) but not in effector or naive T cells. IL-35 is required for the suppressive function of T-reg in vitro and in vivo.